Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human LARGE Full-length ORF (NP_004728.1, 1 a.a. - 756 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00009215P01

Catalog No. 89-950-386


Add to cart

Description

Description

This gene, which is one of the largest in the human genome, encodes a member of the N-acetylglucosaminyltransferase gene family. It encodes a glycosyltransferase which participates in glycosylation of alpha-dystroglycan, and may carry out the synthesis of glycoprotein and glycosphingolipid sugar chains. It may also be involved in the addition of a repeated disaccharide unit. Mutations in this gene cause MDC1D, a novel form of congenital muscular dystrophy with severe mental retardation and abnormal glycosylation of alpha-dystroglycan. Alternative splicing of this gene results in two transcript variants that encode the same protein. [provided by RefSeq]

Sequence: MLGICRGRRKFLAASLSLLCIPAITWIYLFSGSFEDGKPVSLSPLESQAHSPRYTASSQRERESLEVRMREVEEENRALRRQLSLAQGRAPSHRRGNHSKTYSMEEGTGDSENLRAGIVAGNSSECGQQPVVEKCETIHVAIVCAGYNASRDVVTLVKSVLFHRRNPLHFHLIADSIAEQILATLFQTWMVPAVRVDFYNADELKSEVSWIPNKHYSGIYGLMKLVLTKTLPANLERVIVLDTDITFATDIAELWAVFHKFKGQQVLGLVENQSDWYLGNLWKNHRPWPALGRGYNTGVILLLLDKLRKMKWEQMWRLTAERELMGMLSTSLADQDIFNAVIKQNPFLVYQLPCFWNVQLSDHTRSEQCYRDVSDLKVIHWNSPKKLRVKNKHVEFFRNLYLTFLEYDGNLLRRELFGCPSEADVNSENLQKQLSELDEDDLCYEFRRERFTVHRTHLYFLHYEYEPAADSTDVTLVAQLSMDRLQMLEAICKHWEGPISLALYLSDAEAQQFLRYAQGSEVLMSRHNVGYHIVYKEGQFYPVNLLRNVAMKHISTPYMFLSDIDFLPMYGLYEYLRKSVIQLDLANTKKAMIVPAFETLRYRLSFPKSKAELLSMLDMGTLFTFRYHVWTKGHAPTNFAKWRTATTPYRVEWEADFEPYVVVRRDCPEYDRRFVGFGWNKVAHIMELDVQEYEFIVLPNAYMIHMPHAPSFDITKFRSNKQYRICLKTLKEEFQQDMSRRYGFAALKYLTAENNS
Specifications

Specifications

wheat germ expression system
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
114.5kDa
Glutathione Sepharose 4 Fast Flow
2 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
KIAA0609|MDC1D
LARGE
Yes
NP_004728.1
Liquid
9215
LARGE (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MLGICRGRRKFLAASLSLLCIPAITWIYLFSGSFEDGKPVSLSPLESQAHSPRYTASSQRERESLEVRMREVEEENRALRRQLSLAQGRAPSHRRGNHSKTYSMEEGTGDSENLRAGIVAGNSSECGQQPVVEKCETIHVAIVCAGYNASRDVVTLVKSVLFHRRNPLHFHLIADSIAEQILATLFQTWMVPAVRVDFYNADELKSEVSWIPNKHYSGIYGLMKLVLTKTLPANLERVIVLDTDITFATDIAELWAVFHKFKGQQVLGLVENQSDWYLGNLWKNHRPWPALGRGYNTGVILLLLDKLRKMKWEQMWRLTAERELMGMLSTSLADQDIFNAVIKQNPFLVYQLPCFWNVQLSDHTRSEQCYRDVSDLKVIHWNSPKKLRVKNKHVEFFRNLYLTFLEYDGNLLRRELFGCPSEADVNSENLQKQLSELDEDDLCYEFRRERFTVHRTHLYFLHYEYEPAADSTDVTLVAQLSMDRLQMLEAICKHWEGPISLALYLSDAEAQQFLRYAQGSEVLMSRHNVGYHIVYKEGQFYPVNLLRNVAMKHISTPYMFLSDIDFLPMYGLYEYLRKSVIQLDLANTKKAMIVPAFETLRYRLSFPKSKAELLSMLDMGTLFTFRYHVWTKGHAPTNFAKWRTATTPYRVEWEADFEPYVVVRRDCPEYDRRFVGFGWNKVAHIMELDVQEYEFIVLPNAYMIHMPHAPSFDITKFRSNKQYRICLKTLKEEFQQDMSRRYGFAALKYLTAENNS
RUO
LARGE
Wheat Germ (in vitro)
GST
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit