Learn More
Abnova™ Human LARP2 Full-length ORF (AAH30516.1, 1 a.a. - 201 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Supplier: Abnova™ H00055132P01L
Description
This gene encodes a protein containing domains found in the La related protein of Drosophila melanogaster. La motif-containing proteins are thought to be RNA-binding proteins, where the La motif and adjacent amino acids fold into an RNA recognition motif. The La motif is also found in proteins unrelated to the La protein. Alternative splicing has been observed at this locus and three variants, encoding distinct isoforms, are described. Additional splice variation has been identified but the full-length nature of these transcripts has not been determined. [provided by RefSeq]
Sequence: MDSRDHGPGTSSVSTSNASPSEGAPLAGSYGCTPHSFPKFQHPSHELLKENGFTQQVYHKYRRRCLSERKRLGIGQSQEMNTLFRFWSFFLRDHFNKKMYEEFRQLAWEDAKENYRYGLECLFRFYSYGLEKKFRREIFQDFQEETKKDYESGQLYGLEKFWAYLKYSQSKTQSIDPKLQEYLCSFKRLEDFRVDEADPIESpecifications
AAH30516.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
50.5kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DKFZp686L13217/MGC117277/MGC75174 | |
LARP2 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
55132 | |
LARP2 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MDSRDHGPGTSSVSTSNASPSEGAPLAGSYGCTPHSFPKFQHPSHELLKENGFTQQVYHKYRRRCLSERKRLGIGQSQEMNTLFRFWSFFLRDHFNKKMYEEFRQLAWEDAKENYRYGLECLFRFYSYGLEKKFRREIFQDFQEETKKDYESGQLYGLEKFWAYLKYSQSKTQSIDPKLQEYLCSFKRLEDFRVDEADPIE | |
RUO | |
LARP2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.