Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abnova™ Human LARS2 Full-length ORF (NP_056155.1, 1 a.a. - 903 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Specifications
Accession Number | NP_056155.1 |
---|---|
For Use With (Application) | Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 23395 |
Molecular Weight (g/mol) | 128.4kDa |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
89-950-400
|
Abnova™
H00023395P01L |
25μg | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||||
89-950-399
|
Abnova™
H00023395P01S |
10μg | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||||
Description
This gene encodes a class 1 aminoacyl-tRNA synthetase, mitochondrial leucyl-tRNA synthetase. Each of the twenty aminoacyl-tRNA synthetases catalyzes the aminoacylation of a specific tRNA or tRNA isoaccepting family with the cognate amino acid. [provided by RefSeq]
Sequence: MASVWQRLGFYASLLKRQLNGGPDVIKWERRVIPGCTRSIYSATGKWTKEYTLQTRKDVEKWWHQRIKEQASKISEADKSKPKFYVLSMFPYPSGKLHMGHVRVYTISDTIARFQKMRGMQVINPMGWDAFGLPAENAAVERNLHPQSWTQSNIKHMRKQLDRLGLCFSWDREITTCLPDYYKWTQYLFIKLYEAGLAYQKEALVNWDPVDQTVLANEQVDEHGCSWRSGAKVEQKYLRQWFIKTTAYAKAMQDALADLPEWYGIKGMQAHWIGDCVGCHLDFTLKVHGQATGEKLTAYTATPEAIYGTSHVAISPSHRLLHGHSSLKEALRMALVPGKDCLTPVMAVNMLTQQEVPVVILAKADLEGSLDSKIGIPSTSSEDTILAQTLGLAYSEVIETLPDGTERLSSSAEFTGMTRQDAFLALTQKARGKRVGGDVTSDKLKDWLISRQRYWGTPIPIVHCPVCGPTPVPLEDLPVTLPNIASFTGKGGPPLAMASEWVNCSCPRCKGAAKRETDTMDTFVDSAWYYFRYTDPHNPHSPFNTAVADYWMPVDLYIGGKEHAVMHLFYARFFSHFCHDQKMVKHREPFHKLLAQGLIKGQTFRLPSGQYLQREEVDLTGSVPVHAKTKEKLEVTWEKMSKSKHNGVDPEEVVEQYGIDTIRLYILFAAPPEKDILWDVKTDALPGVLRWQQRLWTLTTRFIEARASGKSPQPQLLSNKEKAEARKLWEYKNSVISQVTTHFTEDFSLNSAISQLMGLSNALSQASQSVILHSPEFEDALCALMVMAAPLAPHVTSEIWAGLALVPRKLCAHYTWDASVLLQAWPAVDPEFLQQPEVVQMAVLINNKACGKIPVPQQVARDQDKVHEFVLQSELGVRLLQGRSIKKSFLSPRTALINFLVQDSpecifications
NP_056155.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
128.4kDa | |
Glutathione Sepharose 4 Fast Flow | |
Store at −80°C. Aliquot to avoid repeated freezing and thawing. | |
KIAA0028, LEURS, MGC26121 | |
LARS2 | |
Recombinant | |
wheat germ expression system |
Protein Array, ELISA, Western Blot | |
23395 | |
Human LARS2 Full-length ORF (NP_056155.1, 1 a.a. - 903 a.a.) Recombinant Protein with GST-tag at N-terminal | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
RUO | |
LARS2 | |
Wheat Germ (in vitro) | |
GST Tag | |
Liquid |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title