Learn More
Abnova™ Human LASS1 Full-length ORF (AAH84582.1, 1 a.a. - 239 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Supplier: Abnova™ H00010715P01L
Description
This gene encodes a member of the bone morphogenetic protein (BMP) family and the TGF-beta superfamily. This group of proteins is characterized by a polybasic proteolytic processing site that is cleaved to produce a mature protein containing seven conserved cysteine residues. Members of this family are regulators of cell growth and differentiation in both embryonic and adult tissues. Studies in yeast suggest that the encoded protein is involved in aging. This protein is transcribed from a monocistronic mRNA as well as a bicistronic mRNA, which also encodes growth differentiation factor 1. [provided by RefSeq]
Sequence: MPESAWKFLFYLGSWSYSAYLLFGTDYPFFHDPPSVFYDWTPGMAVPRDIAAAYLLQGSFYGHSIYATLYMDTWRKDSVVMLLHHVVTLILIVSSYAFRYHNVGILVLFLHDISDVQLEFTKLNIYFKSRGGSYHRLHALAADLGCLSFGFSWFWFRLYWFPLKVLYATSHCSLRTVPDIPFYFFFNALLLLLTLMNLYWFLYIVAFAAKVLTGQVHELKDLREYDTAEAQSLKPSKAESpecifications
AAH84582.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
54.3kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CerS1/LAG1/MGC90349/UOG1 | |
LASS1 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
10715 | |
LASS1 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MPESAWKFLFYLGSWSYSAYLLFGTDYPFFHDPPSVFYDWTPGMAVPRDIAAAYLLQGSFYGHSIYATLYMDTWRKDSVVMLLHHVVTLILIVSSYAFRYHNVGILVLFLHDISDVQLEFTKLNIYFKSRGGSYHRLHALAADLGCLSFGFSWFWFRLYWFPLKVLYATSHCSLRTVPDIPFYFFFNALLLLLTLMNLYWFLYIVAFAAKVLTGQVHELKDLREYDTAEAQSLKPSKAE | |
RUO | |
LASS1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.