Learn More
Abnova™ Human LATS1 (NP_004681.1, 589 a.a. - 1130 a.a.) Partial Recombinant Protein with GST-tag expressed in baculovirus infected Sf21 cells
Used for Func, SDS-PAGE
Supplier: Abnova™ P5573
Description
The protein encoded by this gene is a putative serine/threonine kinase that localizes to the mitotic apparatus and complexes with cell cycle controller CDC2 kinase in early mitosis. The protein is phosphorylated in a cell-cycle dependent manner, with late prophase phosphorylation remaining through metaphase. The N-terminal region of the protein binds CDC2 to form a complex showing reduced H1 histone kinase activity, indicating a role as a negative regulator of CDC2/cyclin A. In addition, the C-terminal kinase domain binds to its own N-terminal region, suggesting potential negative regulation through interference with complex formation via intramolecular binding. Biochemical and genetic data suggest a role as a tumor suppressor. This is supported by studies in knockout mice showing development of soft-tissue sarcomas, ovarian stromal cell tumors and a high sensitivity to carcinogenic treatments. [provided by RefSeq]
Specifications
NP_004681.1 | |
Liquid | |
90kDa | |
Baculovirus infected insect cell (Sf21) expression system | |
Loading 1 ug protein in SDS-PAGE | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
WARTS/wts | |
LATS1 | |
Baculovirus | |
GST | |
Liquid |
Functional Study, SDS-PAGE | |
9113 | |
LATS1 (Human) Recombinant Protein | |
Glutathione sepharose chromatography | |
5 ug | |
RUO | |
LATS1 | |
The activity was measured by off-chip mobility shift assay. The enzyme was incubated with fluorescence-labeled substrate and Mg(or Mn)/ATP. The phosphorylated and unphosphorylated substrates were separated and detected by LabChip 3000. Substrate: Srctide. ATP: 1000 uM | |
Recombinant | |
Baculovirus infected insect cell (Sf21) expression system | |
66% by SDS-PAGE/CBB staining |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.