Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human LBR Full-length ORF (NP_002287.2, 1 a.a. - 615 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00003930P01L

 View more versions of this product

Catalog No. 89-950-439

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

The protein encoded by this gene belongs to the ERG4/ERG24 family. It localized in the nuclear envelope inner membrane and anchors the lamina and the heterochromatin to the membrane. It may mediate interaction between chromatin and lamin B. Mutations of this gene has been associated with autosomal recessive HEM/Greenberg skeletal dysplasia. Alternative splicing occurs at this locus and two transcript variants encoding the same protein have been identified. [provided by RefSeq]

Sequence: MPSRKFADGEVVRGRWPGSSLYYEVEILSHDSTSQLYTVKYKDGTELELKENDIKPLTSFRQRKGGSTSSSPSRRRGSRSRSRSRSPGRPPKSARRSASASHQADIKEARREVEVKLTPLILKPFGNSISRYNGEPEHIERNDAPHKNTQEKFSLSQESSYIATQYSLRPRREEVKLKEIDSKEEKYVAKELAVRTFEVTPIRAKDLEFGGVPGVFLIMFGLPVFLFLLLLMCKQKDPSLLNFPPPLPALYELWETRVFGVYLLWFLIQVLFYLLPIGKVVEGTPLIDGRRLKYRLNGFYAFILTSAVIGTSLFQGVEFHYVYSHFLQFALAATVFCVVLSVYLYMRSLKAPRNDLSPASSGNAVYDFFIGRELNPRIGTFDLKYFCELRPGLIGWVVINLVMLLAEMKIQDRAVPSLAMILVNSFQLLYVVDALWNEEALLTTMDIIHDGFGFMLAFGDLVWVPFIYSFQAFYLVSHPNEVSWPMASLIIVLKLCGYVIFRGANSQKNAFRKNPSDPKLAHLKTIHTSTGKNLLVSGWWGFVRHPNYLGDLIMALAWSLPCGFNHILPYFYIIYFTMLLVHREARDEYHCKKKYGVAWEKYCQRVPYRIFPYIY
Specifications

Specifications

wheat germ expression system
AP, Array, ELISA, WB-Re
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
97.1kDa
Glutathione Sepharose 4 Fast Flow
25μg
RUO
LBR
Wheat Germ (in vitro)
GST Tag
NP_002287.2
Liquid
3930
Human LBR Full-length ORF (NP_002287.2, 1 a.a. - 615 a.a.) Recombinant Protein with GST-tag at N-terminal
12.5% SDS-PAGE Stained with Coomassie Blue.
Store at −80°C. Aliquot to avoid repeated freezing and thawing.
DHCR14B, FLJ43126, LMN2R, MGC9041, PHA
LBR
Yes
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit