Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human LCP2 Full-length ORF (AAH16618, 1 a.a. - 533 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00003937P01L

 View more versions of this product

Catalog No. 89-950-480


Add to cart

Description

Description

SLP-76 was originally identified as a substrate of the ZAP-70 protein tyrosine kinase following T cell receptor (TCR) ligation in the leukemic T cell line Jurkat. The SLP-76 locus has been localized to human chromosome 5q33 and the gene structure has been partially characterized in mice. The human and murine cDNAs both encode 533 amino acid proteins that are 72% identical and comprised of three modular domains. The NH2-terminus contains an acidic region that includes a PEST domain and several tyrosine residues which are phosphorylated following TCR ligation. SLP-76 also contains a central proline-rich domain and a COOH-terminal SH2 domain. A number of additional proteins have been identified that associate with SLP-76 both constitutively and inducibly following receptor ligation, supporting the notion that SLP-76 functions as an adaptor or scaffold protein. Studies using SLP-76 deficient T cell lines or mice have provided strong evidence that SLP-76 plays a positive role in promoting T cell development and activation as well as mast cell and platelet function. [provided by RefSeq]

Sequence: MALRNVPFRSEVLGWDPDSLADYFKKLNYKDCEKAVKKYHIDGARFLNLTENDIQKFPKLRVPILSKLSQEINKNEERRSIFTRKPQVPRFPEETESHEEDNGGWSSFEEDDYESPNDDQDGEDDGDYESPNEEEEAPVEDDADYEPPPSNDEEALQNSILPAKPFPNSNSMYIDRPPSGKTPQQPPVPPQRPMAALPPPPAGRNHSPLPPPQTNHEEPSRSRNHKTAKLPAPSIDRSTKPPLDRSLAPFDREPFTLGKKPPFSDKPSIPAGRSLGEHLPKIQKPPLPPTTERHERSSPLPGKKPPVPKHGWGPDRRENDEDDVHQRPLPQPALLPMSSNTFPSRSTKPSPMNPLPSSHMPGAFSESNSSFPQSASLPPYFSQGPSNRPPIRAEGRNFPLPLPNKPRPPSPAEEENSLNEEWYVSYITRPEAEAALRKINQDGTFLVRDSSKKTTTNPYVLMVLYKDKVYNIQIRYQKESQVYLLGTGLRGKEDFLSVSDIIDYFRKMPLLLIDGKNRGSRYQCTLTHAAGYP
Specifications

Specifications

AAH16618
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
84.37kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
SLP-76|SLP76
LCP2
Yes
wheat germ expression system
Liquid
3937
LCP2 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MALRNVPFRSEVLGWDPDSLADYFKKLNYKDCEKAVKKYHIDGARFLNLTENDIQKFPKLRVPILSKLSQEINKNEERRSIFTRKPQVPRFPEETESHEEDNGGWSSFEEDDYESPNDDQDGEDDGDYESPNEEEEAPVEDDADYEPPPSNDEEALQNSILPAKPFPNSNSMYIDRPPSGKTPQQPPVPPQRPMAALPPPPAGRNHSPLPPPQTNHEEPSRSRNHKTAKLPAPSIDRSTKPPLDRSLAPFDREPFTLGKKPPFSDKPSIPAGRSLGEHLPKIQKPPLPPTTERHERSSPLPGKKPPVPKHGWGPDRRENDEDDVHQRPLPQPALLPMSSNTFPSRSTKPSPMNPLPSSHMPGAFSESNSSFPQSASLPPYFSQGPSNRPPIRAEGRNFPLPLPNKPRPPSPAEEENSLNEEWYVSYITRPEAEAALRKINQDGTFLVRDSSKKTTTNPYVLMVLYKDKVYNIQIRYQKESQVYLLGTGLRGKEDFLSVSDIIDYFRKMPLLLIDGKNRGSRYQCTLTHAAGYP
RUO
LCP2
Wheat Germ (in vitro)
GST
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit