Learn More
Kalen Biomedical LLCSupplier Diversity Partner HUMAN LDL LOW DENSITY LIP 10MG DFS Item
Manufacturer:  Kalen Biomedical LLCSupplier Diversity Partner 7702001
Human Low Density Lipoprotein (d=1.019 - 1.063 g/mL) is isolated from plasma. The product is provided in PBS with EDTA as a preservative. The protein concentration is 5 mg/mL and the endotoxin level is less than 0.5 EU /mg of protein.
Catalog No. NC0100146