Learn More
Abnova™ Human LDOC1 Partial ORF (NP_036449, 19 a.a. - 120 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Supplier: Abnova™ H00023641Q01S
Description
The protein encoded by this gene contains a leucine zipper-like motif and a proline-rich region that shares marked similarity with an SH3-binding domain. The protein localizes to the nucleus and is down-regulated in some cancer cell lines. It is thought to regulate the transcriptional response mediated by the nuclear factor kappa B (NF-kappaB). The gene has been proposed as a tumor suppressor gene whose protein product may have an important role in the development and/or progression of some cancers. [provided by RefSeq]
Sequence: LSIENSQLMEQLRLLVCERASLLRQVRPPSCPVPFPETFNGESSRLPEFIVQTASYMLVNENRFCNDAMKVAFLISLLTGEAEEWVVPYIEMDSPILGDYRASpecifications
NP_036449 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.96kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
LSIENSQLMEQLRLLVCERASLLRQVRPPSCPVPFPETFNGESSRLPEFIVQTASYMLVNENRFCNDAMKVAFLISLLTGEAEEWVVPYIEMDSPILGDYRA | |
RUO | |
LDOC1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
23641 | |
LDOC1 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
BCUR1/Mar7/Mart7 | |
LDOC1 | |
Recombinant | |
wheat germ expression system |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.