Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human LEPR Full-length ORF (AAI60009.1) Recombinant Protein without tag

Used for AP, Func, Screening

Manufacturer:  Abnova™ H00003953G01

Catalog No. 89-950-558


Add to cart

Description

Description

Leptin (LEP; MIM 164160), an adipocyte-specific hormone that regulates adipose-tissue mass through hypothalamic effects on satiety and energy expenditure, acts through the leptin receptor (LEPR), a single-transmembrane-domain receptor of the cytokine receptor family.[supplied by OMIM]

Sequence: MICQKFCVVLLHWEFIYVITAFNLSYPITPWRFKLSCMPPNSTYDYFLLPAGLSKNTSNSNGHYETAVEPKFNSSGTHFSNLSKTTFHCCFRSEQDRNCSLCADNIEGKTFVSTVNSLVFQQIDANWNIQCWLKGDLKLFICYVESLFKNLFRNYNYKVHLLYVLPEVLEDSPLVPQKGSFQMVHCNCSVHECCECLVPVPTAKLNDTLLMCLKITSGGVIFQSPLMSVQPINMVKPDPPLGLHMEITDDGNLKISWSSPPLVPFPLQYQVKYSENSTTVIREADKIVSATSLLVDSILPGSSYEVQVRGKRLDGPGIWSDWSTPRVFTTQDVIYFPPKILTSVGSNVSFHCIYKKENKIVPSKEIVWWMNLAEKIPQSQYDVVSDHVSKVTFFNLNETKPRGKFTYDAVYCCNEHECHHRYAELYVIDVNINISCETDGYLTKMTCRWSTSTIQSLAESTLQLRYHRSSLYCSDIPSIHPISEPKDCYLQSDGFYECIFQPIFLLSGYTMWIRINHSLGSLDSPPTCVLPDSVVKPLPPSSVKAEITINIGLLKISWEKPVFPENNLQFQIRYGLSGKEVQWKMYEVYDAKSKSVSLPVPDLCAVYAVQVRCKRLDGLGYWSNWSNPAYTVVMDIKVPMRGPEFWRIINGDTMKKEKNVTLLWKPLMKNDSLCSVQRYVINHHTSCNGTWSEDVGNHTKFTFLWTEQAHTVTVLAINSIGASVANFNLTFSWPMSKVNIVQSLSAYPLNSSCVIVSWILSPSDYKLMYFIIEWKNLNEDGEIKWLRISSSVKKYYIHDHFIPIEKYQFSLYPIFMEGVGKPKIINSFTQDDIEKHQSDAGLYVIVPVIISSSILLLGTLLISHQRMKKLFWEDVPNPKNCSWAQGLNFQKPETFEHLFIKHTASVTCGPLLLEPETISEDISVDTSWKNKDEMMPTTVVSLLSTTDLEKGSVCISDQFNSVNFSEAEGTEVTYEDESQRQPFVKYATLISNSKPSETGEEQGLINSSVTKCFSSKNSPLKDSFSNSSWEIEAQAFFILSDQHPNIISPHLTFSEGLDELLKLEGNFPEENNDKKSIYYLGVTSIKKRESGVLLTDKSRVSCPFPAPCLFTDIRVLQDSCSHFVENNINLGTSSKKTFASYMPQFQTCSTQTHKIMENKMCDLTV
Specifications

Specifications

AAI60009.1
Antibody Production, Compound Screening, Functional Study
Liquid
128.15kDa
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
CD295|OBR
LEPR
Yes
wheat germ expression system with proprietary liposome technology
Liquid
3953
LEPR (Human) Recombinant Protein
MICQKFCVVLLHWEFIYVITAFNLSYPITPWRFKLSCMPPNSTYDYFLLPAGLSKNTSNSNGHYETAVEPKFNSSGTHFSNLSKTTFHCCFRSEQDRNCSLCADNIEGKTFVSTVNSLVFQQIDANWNIQCWLKGDLKLFICYVESLFKNLFRNYNYKVHLLYVLPEVLEDSPLVPQKGSFQMVHCNCSVHECCECLVPVPTAKLNDTLLMCLKITSGGVIFQSPLMSVQPINMVKPDPPLGLHMEITDDGNLKISWSSPPLVPFPLQYQVKYSENSTTVIREADKIVSATSLLVDSILPGSSYEVQVRGKRLDGPGIWSDWSTPRVFTTQDVIYFPPKILTSVGSNVSFHCIYKKENKIVPSKEIVWWMNLAEKIPQSQYDVVSDHVSKVTFFNLNETKPRGKFTYDAVYCCNEHECHHRYAELYVIDVNINISCETDGYLTKMTCRWSTSTIQSLAESTLQLRYHRSSLYCSDIPSIHPISEPKDCYLQSDGFYECIFQPIFLLSGYTMWIRINHSLGSLDSPPTCVLPDSVVKPLPPSSVKAEITINIGLLKISWEKPVFPENNLQFQIRYGLSGKEVQWKMYEVYDAKSKSVSLPVPDLCAVYAVQVRCKRLDGLGYWSNWSNPAYTVVMDIKVPMRGPEFWRIINGDTMKKEKNVTLLWKPLMKNDSLCSVQRYVINHHTSCNGTWSEDVGNHTKFTFLWTEQAHTVTVLAINSIGASVANFNLTFSWPMSKVNIVQSLSAYPLNSSCVIVSWILSPSDYKLMYFIIEWKNLNEDGEIKWLRISSSVKKYYIHDHFIPIEKYQFSLYPIFMEGVGKPKIINSFTQDDIEKHQSDAGLYVIVPVIISSSILLLGTLLISHQRMKKLFWEDVPNPKNCSWAQGLNFQKPETFEHLFIKHTASVTCGPLLLEPETISEDISVDTSWKNKDEMMPTTVVSLLSTTDLEKGSVCISDQFNSVNFSEAEGTEVTYEDESQRQPFVKYATLISNSKPSETGEEQGLINSSVTKCFSSKNSPLKDSFSNSSWEIEAQAFFILSDQHPNIISPHLTFSEGLDELLKLEGNFPEENNDKKSIYYLGVTSIKKRESGVLLTDKSRVSCPFPAPCLFTDIRVLQDSCSHFVENNINLGTSSKKTFASYMPQFQTCSTQTHKIMENKMCDLTV
RUO
LEPR
Wheat Germ (in vitro)
None
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit