Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Leptin/OB Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 2 publications
Supplier: R&D Systems AF398
This item is not returnable.
View return policy
Description
Leptin/OB Polyclonal specifically detects Leptin/OB in Human samples. It is validated for Western Blot, Immunohistochemistry, Neutralization.Specifications
Leptin/OB | |
Polyclonal | |
Unconjugated | |
Q6NT58 | |
LEP | |
E. coli-derived recombinant human Leptin/OB Val22-Cys167 Accession # Q6NT58 | |
100 μg | |
Primary | |
Detects human Leptin/OB in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 25% cross-reactivity with recombinant mouse Leptin/OB is observed. | |
Human | |
IgG |
Western Blot, Immunohistochemistry, Neutralization | |
LYOPH | |
Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL, Neutralization 0.5-3.0 ug/mL | |
FLJ94114, LEP, leptin, leptin (murine obesity homolog), leptin (obesity homolog, mouse), OB, Obese protein, obese, mouse, homolog of, Obesity factor, OBOBS | |
Goat | |
Affinity Purified | |
RUO | |
3952 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction