Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Leptin/OB Biotinylated Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 7 publications
Supplier: R&D Systems BAM398
This item is not returnable.
View return policy
Description
Leptin/OB Monoclonal specifically detects Leptin/OB in Human samples. It is validated for ELISA Detection (Matched Antibody Pair).Specifications
Leptin/OB | |
Monoclonal | |
LYOPH | |
ELISA Detection (Matched Antibody Pair) 0.5-2.0 ug/mL | |
P41159 | |
LEP | |
E. coli-derived recombinant human Leptin Val22-Cys167 Accession # P41159 | |
250 μg | |
Primary | |
Detects human Leptin/OB in ELISAs and Western blots. In ELISAs, this antibody shows less than 0.1% cross-reactivity with recombinant mouse (rm) and rrLeptin. In Western blots, this antibody shows no cross-reactivity with rmLeptin. | |
Human | |
Purified |
ELISA | |
44804 | |
Biotin | |
Lyophilized from a 0.2 μm filtered solution in PBS with BSA as a carrier protein. with No Preservative | |
FLJ94114, LEP, leptin, leptin (murine obesity homolog), leptin (obesity homolog, mouse), OB, Obese protein, obese, mouse, homolog of, Obesity factor, OBOBS | |
Mouse | |
Protein A or G purified from ascites | |
RUO | |
3952 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction