Learn More
BioworldSupplier Diversity Partner HUMAN LEPTIN TRIPLE ANTAGON DFS Item
Manufacturer:  BioworldSupplier Diversity Partner 220604041
HUMAN LEPTIN TRIPLE ANTAGONIST (RECOMBINANT): Recombinant human leptin (hLEP) antagonist (triple mutant) Source: E.Coli Background: Leptin inhibits food intake and stimulates energy expenditure. Leptin also has thermogenic actions an...
Catalog No. 50-253-501