Learn More
Abnova™ Human LGALS1 (P09382) Recombinant Protein
Used for Func, SDS-PAGE
Manufacturer:  Abnova™ P4390
Catalog No. 89-950-572
Add to cart
Description
Description
The galectins are a family of beta-galactoside-binding proteins implicated in modulating cell-cell and cell-matrix interactions. This gene product may act as an autocrine negative growth factor that regulates cell proliferation. [provided by RefSeq]
Sequence: ACGLVASNLNLKPGECLRVRGEVAPDAKSFVLNLGKDSNNLCLHFNPRFNAHGDANTIVCNSKDGGAWGTEQREAVFPFQPGSVAEVCITFDQANLTVKLPDGYEFKFPNRLNLEAINYMAADGDFKIKCVAFD
Specifications
Specifications
P09382
Functional Study, Sodium Dodecyl Sulfate–Polyacrylamide Gel Electrophoresis
Lyophilized
14.5kDa
Escherichia coli expression system
50 ug
Store at -20°C on dry atmosphere. After reconstitution with sterilized water, store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
<0.1 EU/μg
LGALS1
The activity is determined by the ability to induce chemotaxis of human THP-1 cells and is detectable starting at 100ng/mL.
Yes
Escherichia coli expression system
Lyophilized
3956
LGALS1 (Human) Recombinant Protein
1 ug/lane in 4-20% Tris-Glycine gel Stained with Coomassie Blue
MACGLVASNLNLKPGECLRVRGEVAPDAKSFVLNLGKDSNNLCLHFNPRFNAHGDANTIVCNSKDGGAWGTEQREAVFPFQPGSVAEVCITFDQANLTVKLPDGYEFKFPNRLNLEAINYMAADGDFKIKCVAFD
RUO
DKFZp686E23103|GAL1|GBP
LGALS1
Escherichia coli
None