Learn More
Abnova™ Human LGALS14 Partial ORF (NP_064514.1, 73 a.a. - 139 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
$499.00 - $769.00
Specifications
Accession Number | NP_064514.1 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 56891 |
Molecular Weight (g/mol) | 33.11kDa |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
89-950-579
|
Abnova™
H00056891Q01L |
25 ug |
Each for $769.00
|
|
|||||
89-950-578
|
Abnova™
H00056891Q01S |
10 ug |
Each for $499.00
|
|
|||||
Description
This gene is predominantly expressed in placenta. The encoded protein belongs to the galectin (galaptin/S-lectin) family. The members of galectin family contain one or two carbohydrate recognition domains, which can bind beta-galactoside. Two alternatively spliced transcript variants encoding distinct isoforms have been observed. [provided by RefSeq]
Sequence: RYEEKCYYLPFEDGKPFELCIYVRHKEYKVMVNGQRIYNFAHRFPPASVKMLQVLRDISLTRVLISDSpecifications
NP_064514.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
33.11kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CLC2/MGC22235/PPL13 | |
LGALS14 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
56891 | |
LGALS14 (Human) Recombinant Protein (Q01) | |
RYEEKCYYLPFEDGKPFELCIYVRHKEYKVMVNGQRIYNFAHRFPPASVKMLQVLRDISLTRVLISD | |
RUO | |
LGALS14 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.