Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abnova™ Human LGALS2 (NP_006489, 1 a.a. - 132 a.a.) Full-length Recombinant Protein with His tag
Used for SDS-PAGE
Supplier: Abnova™ P3410
Description
The protein encoded by this gene is a soluble beta-galactoside binding lectin. The encoded protein is found as a homodimer and can bind to lymphotoxin-alpha. A single nucleotide polymorphism in an intron of this gene can alter the transcriptional level of the protein, with a resultant increased risk of myocardial infarction. [provided by RefSeq]
Sequence: MGSSHHHHHHSSGLVPRGSHMTGELEVKNMDMKPGSTLKITGSIADGTDGFVINLGQGTDKLNLHFNPRFSESTIVCNSLDGSNWGQEQREDHLCFSPGSEVKFTVTFESDKFKVKLPDGHELTFPNRLGHSHLSYLSVRGGFNMSSFKLKESpecifications
NP_006489 | |
SDS-PAGE | |
3957 | |
LGALS2 (Human) Recombinant Protein | |
Conventional Chromatography | |
100 ug | |
Store at 4°C for 1-2 weeks. For long term storage store at -20°C or -80°C. Aliquot to avoid repeated freezing and thawing. | |
HL14/MGC75071 | |
LGALS2 | |
Recombinant | |
Escherichia coli expression system | |
>95% by SDS-PAGE |
0.5 mg/mL | |
Liquid | |
16.8kDa | |
Escherichia coli expression system | |
Loading 3 ug protein in 15% SDS-PAGE | |
MGSSHHHHHHSSGLVPRGSHMTGELEVKNMDMKPGSTLKITGSIADGTDGFVINLGQGTDKLNLHFNPRFSESTIVCNSLDGSNWGQEQREDHLCFSPGSEVKFTVTFESDKFKVKLPDGHELTFPNRLGHSHLSYLSVRGGFNMSSFKLKE | |
RUO | |
LGALS2 | |
E. coli | |
His | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction