Learn More
Abnova™ Human LGALS3 (Q61BA7) Recombinant Protein
Used for Func, SDS-PAGE
Manufacturer:  Abnova™ P4391
Catalog No. 89-950-586
Add to cart
Description
Description
Sequence: ADNFSLHDALSGSGNPNPQGWPGAWGNQPAGAGGYPGASYPGAYPGQAPPGAYPGQAPPGAYHGAPGAYPGAPAPGVYPGPPSGPGAYPSSGQPSAPGAYPATGPYGAPAGPLIVPYNLPLPGGVVPRMLITILGTVKPNANRIALDFQRGNDVAFHFNPRFNENNRRVIVCNTKLDNNWGREERQSVFPFESGKPFKIQVLVEPDHFKVAVNDAHLLQYNHRVKKLNEISK
Specifications
Specifications
Q61BA7
Functional Study, Sodium Dodecyl Sulfate–Polyacrylamide Gel Electrophoresis
Lyophilized
26kDa
Escherichia coli expression system
50 ug
Store at -20°C on dry atmosphere. After reconstitution with sterilized water, store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
<0.1 EU/μg
LGALS3
The activity is determined by the ability to induce chemotaxis of human PBMCs and is detectable starting at 1μg/mL.
Yes
Escherichia coli expression system
Lyophilized
3958
LGALS3 (Human) Recombinant Protein
1 ug/lane in 4-20% Tris-Glycine gel Stained with Coomassie Blue
MADNFSLHDALSGSGNPNPQGWPGAWGNQPAGAGGYPGASYPGAYPGQAPPGAYPGQAPPGAYHGAPGAYPGAPAPGVYPGPPSGPGAYPSSGQPSAPGAYPATGPYGAPAGPLIVPYNLPLPGGVVPRMLITILGTVKPNANRIALDFQRGNDVAFHFNPRFNENNRRVIVCNTKLDNNWGREERQSVFPFESGKPFKIQVLVEPDHFKVAVNDAHLLQYNHRVKKLNEISKLGISGDIDLTSASYTMI
RUO
CBP35|GAL3|GALBP|GALIG|LGALS2|MAC2
LGALS3
Escherichia coli
None