Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abnova™ Human LGALS4 Partial ORF (NP_006140, 51 a.a. - 150 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Supplier: Abnova™ H00003960Q01S
Description
The galectins are a family of beta-galactoside-binding proteins implicated in modulating cell-cell and cell-matrix interactions. The expression of this gene is restricted to small intestine, colon, and rectum, and it is underexpressed in colorectal cancer. [provided by RefSeq]
Sequence: VVGQDPGSDVAFHFNPRFDGWDKVVFNTLQGGKWGSEERKRSMPFKKGAAFELVFIVLAEHYKVVVNGNPFYEYGHRLPLQMVTHLQVDGDLQLQSINFISpecifications
NP_006140 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
VVGQDPGSDVAFHFNPRFDGWDKVVFNTLQGGKWGSEERKRSMPFKKGAAFELVFIVLAEHYKVVVNGNPFYEYGHRLPLQMVTHLQVDGDLQLQSINFI | |
RUO | |
LGALS4 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
3960 | |
LGALS4 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
GAL4/L36LBP | |
LGALS4 | |
Recombinant | |
wheat germ expression system |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction