Learn More
Abnova™ Human LGALS7 (P47929, 136 amino acids) Partial Recombinant Protein
Used for Func, SDS-PAGE
Manufacturer:  Abnova™ P4114
Catalog No. 89-950-592
Add to cart
Description
Description
The galectins are a family of beta-galactoside-binding proteins implicated in modulating cell-cell and cell-matrix interactions. Differential and in situ hybridization studies indicate that this lectin is specifically expressed in keratinocytes and found mainly in stratified squamous epithelium. A duplicate copy of this gene (GeneID:653499) is found adjacent to, but on the opposite strand on chromosome 19. [provided by RefSeq]
Specifications
Specifications
Escherichia coli expression system
Functional Study, Sodium Dodecyl Sulfate–Polyacrylamide Gel Electrophoresis
Lyophilized
15.4kDa
Escherichia coli expression system
SDS-PAGE Stained with Coomassie Blue
Store at -20°C on dry atmosphere. After reconstitution with sterilized water, store at -20°C. Aliquot to avoid repeated freezing and thawing.
<0.1ng/μg
LGALS7
Measured by its ability to agglutinate human red blood cells. The ED50 for the effect is typically 1-7μg/mL.
Yes
P47929
Lyophilized
3963
LGALS7 (Human) Recombinant Protein
≥98% by SDS-PAGE and HPLC
10 ug
RUO
GAL7|LGALS7A
LGALS7
Escherichia coli
None