Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human LHX2 Full-length ORF (NP_004780.3, 1 a.a. - 406 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00009355P01L

 View more versions of this product

Catalog No. 89-950-635


Add to cart

Description

Description

This gene encodes a protein belonging to a large protein family, members of which carry the LIM domain, a unique cysteine-rich zinc-binding domain. The encoded protein may function as a transcriptional regulator. The protein can recapitulate or rescue phenotypes in Drosophila caused by a related protein, suggesting conservation of function during evolution. [provided by RefSeq]

Sequence: MLFHSLSGPEVHGVIDEMDRRAKSEAPAISSAIDRGDTETTMPSISSDRAALCAGCGGKISDRYYLLAVDKQWHMRCLKCCECKLNLESELTCFSKDGSIYCKEDYYRRFSVQRCARCHLGISASEMVMRARDLVYHLNCFTCTTCNKMLTTGDHFGMKDSLVYCRLHFEALLQGEYPAHFNHADVAAAAAAAAAAKSAGLGAAGANPLGLPYYNGVGTVQKGRPRKRKSPGPGADLAAYNAALSCNENDAEHLDRDQPYPSSQKTKRMRTSFKHHQLRTMKSYFAINHNPDAKDLKQLAQKTGLTKRVLQVWFQNARAKFRRNLLRQENTGVDKSTDAALQTGTPSGPASELSNASLSPSSTPTTLTDLTSPTLPTVTSVLTSVPGNLEGHEPHSPSQTTLTNLF
Specifications

Specifications

NP_004780.3
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
70.8kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
LH2|MGC138390|hLhx2
LHX2
Yes
wheat germ expression system
Liquid
9355
LHX2 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MLFHSLSGPEVHGVIDEMDRRAKSEAPAISSAIDRGDTETTMPSISSDRAALCAGCGGKISDRYYLLAVDKQWHMRCLKCCECKLNLESELTCFSKDGSIYCKEDYYRRFSVQRCARCHLGISASEMVMRARDLVYHLNCFTCTTCNKMLTTGDHFGMKDSLVYCRLHFEALLQGEYPAHFNHADVAAAAAAAAAAKSAGLGAAGANPLGLPYYNGVGTVQKGRPRKRKSPGPGADLAAYNAALSCNENDAEHLDRDQPYPSSQKTKRMRTSFKHHQLRTMKSYFAINHNPDAKDLKQLAQKTGLTKRVLQVWFQNARAKFRRNLLRQENTGVDKSTDAALQTGTPSGPASELSNASLSPSSTPTTLTDLTSPTLPTVTSVLTSVPGNLEGHEPHSPSQTTLTNLF
RUO
LHX2
Wheat Germ (in vitro)
GST
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit