Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human LHX2 Partial ORF (NP_004780, 1 a.a. - 76 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

$499.00 - $769.00

Specifications

Accession Number NP_004780
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 9355
Molecular Weight (g/mol) 34.1kDa
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
89-950-632
View Documents
Abnova™
H00009355Q01S
10 ug
Each for $499.00
Add to cart
 
89-950-633
View Documents
Abnova™
H00009355Q01L
25 ug
Each for $769.00
Add to cart
 
Description

Description

This gene encodes a protein belonging to a large protein family, members of which carry the LIM domain, a unique cysteine-rich zinc-binding domain. The encoded protein may function as a transcriptional regulator. The protein can recapitulate or rescue phenotypes in Drosophila caused by a related protein, suggesting conservation of function during evolution. [provided by RefSeq]

Sequence: MLFHSLSGPEVHGVIDEMDRRAKSEAPAISSAIDRGDTETTMPSISSDRAALCAGCGGKISDRYYLLAVDKQWHMR
Specifications

Specifications

NP_004780
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
34.1kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
LH2/MGC138390/hLhx2
LHX2
Recombinant
wheat germ expression system
Antibody Production, ELISA, Protein Array, Western Blot
9355
LHX2 (Human) Recombinant Protein (Q01)
MLFHSLSGPEVHGVIDEMDRRAKSEAPAISSAIDRGDTETTMPSISSDRAALCAGCGGKISDRYYLLAVDKQWHMR
RUO
LHX2
Wheat Germ (in vitro)
GST
Liquid
SDS
missing translation for 'documents'

missing translation for 'documents'

Product Certifications
missing translation for 'provideContentCorrection'

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

missing translation for 'productTitle'