Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human LHX4 Full-length ORF (NP_203129.1, 1 a.a. - 390 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00089884P01

Catalog No. 89-950-640


Add to cart

Description

Description

This gene encodes a member of a large protein family which contains the LIM domain, a unique cysteine-rich zinc-binding domain. The encoded protein may function as a transcriptional regulator and be involved in control of differentiation and development of the pituitary gland. Mutations in this gene are associated with syndromic short stature and pituitary and hindbrain defects. An alternative splice variant has been described but its biological nature has not been determined. [provided by RefSeq]

Sequence: MMQSATVPAEGAVKGLPEMLGVPMQQIPQCAGCNQHILDKFILKVLDRHWHSSCLKCADCQMQLADRCFSRAGSVYCKEDFFKRFGTKCTACQQGIPPTQVVRKAQDFVYHLHCFACIICNRQLATGDEFYLMEDGRLVCKEDYETAKQNDDSEAGAKRPRTTITAKQLETLKNAYKNSPKPARHVREQLSSETGLDMRVVQVWFQNRRAKEKRLKKDAGRHRWGQFYKSVKRSRGSSKQEKESSAEDCGVSDSELSFREDQILSELGHTNRIYGNVGDVTGGQLMNGSFSMDGTGQSYQDLRDGSPYGIPQSPSSISSLPSHAPLLNGLDYTVDSNLGIIAHAGQGVSQTLRAMAGGPTSDISTGSSVGYPDFPTSPGSWLDEMDHPPF
Specifications

Specifications

NP_203129.1
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
69.5kDa
Glutathione Sepharose 4 Fast Flow
2 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Gsh-4|Gsh4
LHX4
Yes
wheat germ expression system
Liquid
89884
LHX4 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MMQSATVPAEGAVKGLPEMLGVPMQQIPQCAGCNQHILDKFILKVLDRHWHSSCLKCADCQMQLADRCFSRAGSVYCKEDFFKRFGTKCTACQQGIPPTQVVRKAQDFVYHLHCFACIICNRQLATGDEFYLMEDGRLVCKEDYETAKQNDDSEAGAKRPRTTITAKQLETLKNAYKNSPKPARHVREQLSSETGLDMRVVQVWFQNRRAKEKRLKKDAGRHRWGQFYKSVKRSRGSSKQEKESSAEDCGVSDSELSFREDQILSELGHTNRIYGNVGDVTGGQLMNGSFSMDGTGQSYQDLRDGSPYGIPQSPSSISSLPSHAPLLNGLDYTVDSNLGIIAHAGQGVSQTLRAMAGGPTSDISTGSSVGYPDFPTSPGSWLDEMDHPPF
RUO
LHX4
Wheat Germ (in vitro)
GST
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit