Learn More
Abnova™ Human LHX6 Partial ORF (NP_055183, 274 a.a. - 363 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Supplier: Abnova™ H00026468Q01L
Description
This gene encodes a member of a large protein family that contains the LIM domain, a unique cysteine-rich zinc-binding domain. The encoded protein may function as a transcriptional regulator and may be involved in the control of differentiation and development of neural and lymphoid cells. Two alternatively spliced transcript variants encoding distinct isoforms have been described for this gene. Alternatively spliced transcript variants have been identified, but their biological validity has not been determined. [provided by RefSeq]
Sequence: HKKHTPQHPVPPSGAPPSRLPSALSDDIHYTPFSSPERARMVTLHGYIESQVQCGQVHCRLPYTAPPVHLKADMDGPLSNRGEKVILFQYSpecifications
NP_055183 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
35.64kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
HKKHTPQHPVPPSGAPPSRLPSALSDDIHYTPFSSPERARMVTLHGYIESQVQCGQVHCRLPYTAPPVHLKADMDGPLSNRGEKVILFQY | |
RUO | |
LHX6 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
26468 | |
LHX6 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
LHX6.1/MGC119542/MGC119544/MGC119545 | |
LHX6 | |
Recombinant | |
wheat germ expression system |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.