Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human LHX9 Full-length ORF (NP_064589.2, 1 a.a. - 397 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00056956P01S

 View more versions of this product

Catalog No. 89-950-649


Add to cart

Description

Description

This gene encodes a member of the LIM homeobox gene family of developmentally expressed transcription factors. The encoded protein contains a homeodomain and two cysteine-rich zinc-binding LIM domains involved in protein-protein interactions. The protein is highly similar to a mouse protein that causes gonadal agenesis when inactivated, suggesting a role in gonadal development. Alternative splicing results in multiple transcript variants. [provided by RefSeq]

Sequence: MEIVGCRAEDNSCPFRPPAMLFHGISGGHIQGIMEEMERRSKTEARLAKGAQLNGRDAGMPPLSPEKPALCAGCGGKISDRYYLLAVDKQWHLRCLKCCECKLALESELTCFAKDGSIYCKEDYYRRFSVQRCARCHLGISASEMVMRARDSVYHLSCFTCSTCNKTLTTGDHFGMKDSLVYCRAHFETLLQGEYPPQLSYTELAAKSGGLALPYFNGTGTVQKGRPRKRKSPALGVDIVNYNSGCNENEADHLDRDQQPYPPSQKTKRMRTSFKHHQLRTMKSYFAINHNPDAKDLKQLAQKTGLTKRVLQVWFQNARAKFRRNLLRQENGGVDKADGTSLPAPPSADSGALTPPGTATTLTDLTNPTITVVTSVTSNMDSHESGSPSQTTLTNLF
Specifications

Specifications

wheat germ expression system
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
70.4kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
LHX9
Wheat Germ (in vitro)
GST
NP_064589.2
Liquid
56956
LHX9 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MEIVGCRAEDNSCPFRPPAMLFHGISGGHIQGIMEEMERRSKTEARLAKGAQLNGRDAGMPPLSPEKPALCAGCGGKISDRYYLLAVDKQWHLRCLKCCECKLALESELTCFAKDGSIYCKEDYYRRFSVQRCARCHLGISASEMVMRARDSVYHLSCFTCSTCNKTLTTGDHFGMKDSLVYCRAHFETLLQGEYPPQLSYTELAAKSGGLALPYFNGTGTVQKGRPRKRKSPALGVDIVNYNSGCNENEADHLDRDQQPYPPSQKTKRMRTSFKHHQLRTMKSYFAINHNPDAKDLKQLAQKTGLTKRVLQVWFQNARAKFRRNLLRQENGGVDKADGTSLPAPPSADSGALTPPGTATTLTDLTNPTITVVTSVTSNMDSHESGSPSQTTLTNLF
RUO
LHX9
Yes
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit