Learn More
Abnova™ Human LHX9 Full-length ORF (NP_064589.2, 1 a.a. - 397 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Supplier: Abnova™ H00056956P01L
Description
This gene encodes a member of the LIM homeobox gene family of developmentally expressed transcription factors. The encoded protein contains a homeodomain and two cysteine-rich zinc-binding LIM domains involved in protein-protein interactions. The protein is highly similar to a mouse protein that causes gonadal agenesis when inactivated, suggesting a role in gonadal development. Alternative splicing results in multiple transcript variants. [provided by RefSeq]
Sequence: MEIVGCRAEDNSCPFRPPAMLFHGISGGHIQGIMEEMERRSKTEARLAKGAQLNGRDAGMPPLSPEKPALCAGCGGKISDRYYLLAVDKQWHLRCLKCCECKLALESELTCFAKDGSIYCKEDYYRRFSVQRCARCHLGISASEMVMRARDSVYHLSCFTCSTCNKTLTTGDHFGMKDSLVYCRAHFETLLQGEYPPQLSYTELAAKSGGLALPYFNGTGTVQKGRPRKRKSPALGVDIVNYNSGCNENEADHLDRDQQPYPPSQKTKRMRTSFKHHQLRTMKSYFAINHNPDAKDLKQLAQKTGLTKRVLQVWFQNARAKFRRNLLRQENGGVDKADGTSLPAPPSADSGALTPPGTATTLTDLTNPTITVVTSVTSNMDSHESGSPSQTTLTNLFSpecifications
NP_064589.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
70.4kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
LHX9 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
56956 | |
LHX9 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MEIVGCRAEDNSCPFRPPAMLFHGISGGHIQGIMEEMERRSKTEARLAKGAQLNGRDAGMPPLSPEKPALCAGCGGKISDRYYLLAVDKQWHLRCLKCCECKLALESELTCFAKDGSIYCKEDYYRRFSVQRCARCHLGISASEMVMRARDSVYHLSCFTCSTCNKTLTTGDHFGMKDSLVYCRAHFETLLQGEYPPQLSYTELAAKSGGLALPYFNGTGTVQKGRPRKRKSPALGVDIVNYNSGCNENEADHLDRDQQPYPPSQKTKRMRTSFKHHQLRTMKSYFAINHNPDAKDLKQLAQKTGLTKRVLQVWFQNARAKFRRNLLRQENGGVDKADGTSLPAPPSADSGALTPPGTATTLTDLTNPTITVVTSVTSNMDSHESGSPSQTTLTNLF | |
RUO | |
LHX9 | |
Recombinant | |
wheat germ expression system |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.