Learn More
Novus Biologicals™ Human LIF ELISA Kit
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture. 
Manufacturer:  Novus Biologicals™ NBP191186
Catalog No. NBP191186
Add to cart
Description
Description
The Human LIF ELISA Kit (Colorimetric) measures human LIF in cell culture supernatant, serum, plasma (edta, citrate, heparin).
Specifications
Specifications
3.15 - 200pg/mL
3976
ELISA Kit
0.7pg/mL
Human (Hu)
LIF
LIF
ELISA
Aluminium pouch(es) with a Microwell Plate coated with monoclonalantibody to human LIF, Biotin-Conjugate anti-human LIF monoclonal antibody, Streptavidin-HRP, Human LIF Standard, 400 pg/mL, Sample Diluent, Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA), Wash Buffer Concentrate 20x (PBS with 1% Tween 20), Substrate Solution (tetramethyl-benzidine), Stop Solution (1M Phosphoric acid), Adhesive Films
96 Tests
Cell culture supernatant, serum, plasma (EDTA, citrate, heparin)
Storage of components varies. See protocol for specific instructions.
Biologically Active Proteins, Cell Cycle and Replication, Stem Cell Lines
CDF, D factor, DIA, differentiation inhibitory activity, differentiation stimulating factor, Differentiation-stimulating factor, Emfilermin, HILDAcholinergic differentiation factor, leukemia inhibitory factor, leukemia inhibitory factor (cholinergic diffe