Description
- Three parts, including calvaria, base of skull, and mandible
- Hand painted, color-coded to show the various structure of the human skull
- Mandible is articulated to demonstrate movement
- Model features realistic details, textures and bony landmarks
- Made of durable and sturdy plastisol polymer. This washable model is easy to clean and will hold up to years of handling
Specifications
Specifications
| Material | Plastic |
| Product Type | Model |