Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human LIFR Partial ORF (NP_002301, 45 a.a. - 154 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00003977Q01S

 View more versions of this product

Catalog No. 89-950-659


Add to cart

Description

Description

This gene encodes a protein that belongs to the type I cytokine receptor family. This protein combines with a high-affinity converter subunit, gp130, to form a receptor complex that mediates the action of the leukemia inhibitory factor, a polyfunctional cytokine that is involved in cellular differentiation, proliferation and survival in the adult and the embryo. Mutations in this gene cause Schwartz-Jampel syndrome type 2, a disease belonging to the group of the bent-bone dysplasias. A translocation that involves the promoter of this gene, t(5;8)(p13;q12) with the pleiomorphic adenoma gene 1, is associated with salivary gland pleiomorphic adenoma, a common type of benign epithelial tumor of the salivary gland. Multiple splice variants encoding the same protein have been found for this gene. [provided by RefSeq]

Sequence: QKKGAPHDLKCVTNNLQVWNCSWKAPSGTGRGTDYEVCIENRSRSCYQLEKTSIKIPALSHGDYEITINSLHDFGSSTSKFTLNEQNVSLIPDTPEILNLSADFSTSTLY
Specifications

Specifications

NP_002301
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
37.84kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
QKKGAPHDLKCVTNNLQVWNCSWKAPSGTGRGTDYEVCIENRSRSCYQLEKTSIKIPALSHGDYEITINSLHDFGSSTSKFTLNEQNVSLIPDTPEILNLSADFSTSTLY
RUO
LIFR
Wheat Germ (in vitro)
GST
wheat germ expression system
Liquid
3977
LIFR (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
CD118|FLJ98106|FLJ99923|LIF-R|SJS2|STWS|SWS
LIFR
Yes
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit