Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human LILRA3 Partial ORF (NP_006856.2, 340 a.a. - 439 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00011026Q01S

 View more versions of this product

Catalog No. 89-950-675


Add to cart

Description

Description

Leukocyte Ig-like receptors (LIRs) are a family of immunoreceptors expressed predominantly on monocytes and B cells and at lower levels on dendritic cells and natural killer (NK) cells. All LIRs in subfamily B have an inhibitory function (see, e.g., LILRB1, MIM 604811). LIRs in subfamily A, with short cytoplasmic domains lacking an immunoreceptor tyrosine-based inhibitory motif (ITIM) and with transmembrane regions containing a charged arginine residue, may initiate stimulatory cascades (see LILRA1, MIM 604810). One member of subfamily A (LILRA3) lacks a transmembrane region and is presumed to be a soluble receptor (Borges et al., 1997 [PubMed 9548455]).[supplied by OMIM]

Sequence: ENVTLLCQSQGGMHTFLLTKEGAADSPLRLKSKRQSHKYQAEFPMSPVTSAHAGTYRCYGSLSSNPYLLTHPSDPLELVVSGAAETLSPPQNKSDSKAGE
Specifications

Specifications

wheat germ expression system
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.74kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
ENVTLLCQSQGGMHTFLLTKEGAADSPLRLKSKRQSHKYQAEFPMSPVTSAHAGTYRCYGSLSSNPYLLTHPSDPLELVVSGAAETLSPPQNKSDSKAGE
RUO
LILRA3
Wheat Germ (in vitro)
GST
NP_006856.2
Liquid
11026
LILRA3 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
CD85E/HM31/HM43/ILT6/LIR-4/LIR4/e3
LILRA3
Yes
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit