Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems™ Recombinant Human LILRB1/CD85j/ILT2 His-tag Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.
Supplier: R&D Systems™ 8989T2050
This item is not returnable.
View return policy
Description
Measured by its binding ability in a functional ELISA. When Recombinant Human LILRB1/CD85j/ILT2 is coated at 2 μg/mL, Recombinant Human Angiopoietin-like Protein 7/ANGPTL7 (Catalog # 914-AN ) binds with a typical ED50 of 0.075-0.45 μg/mL.Specifications
Q8NHL6 | |
Lyophilized from a 0.2 μm filtered solution in PBS. | |
M.W. (Observed): 65-82 kDa, reducing conditions; M.W. (theoretical): 48 kDa | |
Human embryonic kidney cell, HEK293-derived human LILRB1/CD85j/ILT2 protein Human LILRB1 (Gly24-His458) (N-terminus) Accession # Q8NHL6 HHHHHH (C-terminus) | |
Monomer | |
CD85 antigen-like family member J, CD85J, CD85j antigen, Ig-like transcript 2, ILT2, ILT-2, ILT2FLJ37515, Immunoglobulin-like transcript 2, leukocyte Ig-like receptor-1, Leukocyte immunoglobulin-like receptor 1, leukocyte immunoglobulin-like receptor subfamily B member 1, leukocyte immunoglobulin-like receptor subfamily B member 1 soluble isoform, leukocyte immunoglobulin-like receptor, subfamily B (with TM and ITIM domains), LIR1, LIR1LIR-1MIR-7CD85MIR7CD85 antigen, member 1, MIR7, Monocyte/macrophage immunoglobulin-like receptor 7 | |
LILRB1 | |
Unconjugated | |
Recombinant | |
Human embryonic kidney cell, HEK293-derived Human LILRB1 (Gly24-His458) Accession # Q8NHL6, N-terminus; HHHHHH, C-terminus | |
ILT2/CD85j/LILRB1 |
Bioactivity | |
10859 | |
50 μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution. | |
<0.10 EU / 1 μg of the protein by the LAL method. | |
ILT2/CD85j/LILRB1 | |
Measured by its binding ability in a functional ELISA. When Recombinant Human LILRB1/CD85j/ILT2 is coated at 2μg/mL, Recombinant Human Angiopoietin-like Protein 7/ANGPTL7 binds with a typical ED50 of 75-450ng/mL. | |
Human | |
Reconstitute at 100 μg/mL in PBS. | |
>95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction