Learn More
Abnova™ Human LILRB2 Full-length ORF (AAH41708.1) Recombinant Protein without tag
Used for AP, Func, Screening
Supplier: Abnova™ H00010288G01
Description
This gene is a member of the leukocyte immunoglobulin-like receptor (LIR) family, which is found in a gene cluster at chromosomal region 19q13.4. The encoded protein belongs to the subfamily B class of LIR receptors which contain two or four extracellular immunoglobulin domains, a transmembrane domain, and two to four cytoplasmic immunoreceptor tyrosine-based inhibitory motifs (ITIMs). The receptor is expressed on immune cells where it binds to MHC class I molecules on antigen-presenting cells and transduces a negative signal that inhibits stimulation of an immune response. It is thought to control inflammatory responses and cytotoxicity to help focus the immune response and limit autoreactivity. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]
Sequence: MTPALTALLCLGLSLGPRTRVQAGPFPKPTLWAEPGSVISWGSPVTIWCQGSLEAQEYQLDKEGSPEPLDRNNPLEPKNKARFSIPSMTQHHAGRYRCHYYSSAGWSEPSDPLELVMTGFYNKPTLSALPSPVVASGGNMTLRCGSQKGYHHFVLMKEGEHQLPRTLDSQQLHSGGFQALFPVGPVTPSHRRVSpecifications
AAH41708.1 | |
Liquid | |
21.2kDa | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CD85D/ILT4/LILRA6/LIR-2/LIR2/MIR-10/MIR10 | |
LILRB2 | |
Recombinant | |
wheat germ expression system with proprietary liposome technology |
Antibody Production, Compound Screening, Functional Study | |
10288 | |
LILRB2 (Human) Recombinant Protein | |
MTPALTALLCLGLSLGPRTRVQAGPFPKPTLWAEPGSVISWGSPVTIWCQGSLEAQEYQLDKEGSPEPLDRNNPLEPKNKARFSIPSMTQHHAGRYRCHYYSSAGWSEPSDPLELVMTGFYNKPTLSALPSPVVASGGNMTLRCGSQKGYHHFVLMKEGEHQLPRTLDSQQLHSGGFQALFPVGPVTPSHRRV | |
RUO | |
LILRB2 | |
Wheat Germ (in vitro) | |
None | |
Liquid |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.