Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human LIME1 Full-length ORF (AAH17016.1, 1 a.a. - 295 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00054923P01S

 View more versions of this product

Catalog No. 89-950-693


Add to cart

Description

Description

LIME1 is a raft-associated transmembrane adaptor phosphoprotein that is preferentially expressed in hemopoietic cells, particularly T cells (Brdickova et al., 2003 [PubMed 14610046]; Hur et al., 2003 [PubMed 14610044]).[supplied by OMIM]

Sequence: MGLPVSWAPPALWVLGCCALLLSLWALCTACRRPEDAVAPRKRARRQRARLQGSATAAEASLLRRTHLCSLSKSDTRLHELHRGPRSSRALRPASMDLLRPHWLEVSRDITGPQAAPSAFPHQELPRALPAAAATAGCAGLEATYSNVGLAALPGVSLAASPVVAEYARVQKRKGTHRSPQEPQQGKTEVTPAAQVDVLYSRVCKPKRRDPGPTTDPLDPKGQGAILALAGDLAYQTLPLRALDVDSGPLENVYESIRELGDPAGRSSTCGAGTPPASSCPSLGRGWRPLPASLP
Specifications

Specifications

AAH17016.1
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
57.7kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FLJ20406|LIME|LP8067|dJ583P15.4
LIME1
Yes
wheat germ expression system
Liquid
54923
LIME1 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MGLPVSWAPPALWVLGCCALLLSLWALCTACRRPEDAVAPRKRARRQRARLQGSATAAEASLLRRTHLCSLSKSDTRLHELHRGPRSSRALRPASMDLLRPHWLEVSRDITGPQAAPSAFPHQELPRALPAAAATAGCAGLEATYSNVGLAALPGVSLAASPVVAEYARVQKRKGTHRSPQEPQQGKTEVTPAAQVDVLYSRVCKPKRRDPGPTTDPLDPKGQGAILALAGDLAYQTLPLRALDVDSGPLENVYESIRELGDPAGRSSTCGAGTPPASSCPSLGRGWRPLPASLP
RUO
LIME1
Wheat Germ (in vitro)
GST
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit