Learn More
Abnova™ Human LIN28 (Q9H9Z2) Full-length Recombinant Protein
Used for Func, SDS-PAGE
Manufacturer:  Abnova™ P6110
Catalog No. 89-950-713
Add to cart
Description
Description
Sequence: GPSVSNQQFAGGCAKAAEEAPEEAPEDAARAADEPQLLHGAGICKWFNVRMGFGFLSMTARAGVALDPPVDVFVHQSKLHMEGFRSLKEGEAVEFTFKKSAKGLESIRVTGPGGVFCIGSERRPKGKSMQKRRSKGDRCYNCGGLDHHAKECKLPPQPKKCHFCQSISHMVASCPLKAQQGP
Specifications
Specifications
Q9H9Z2
Functional Study, Sodium Dodecyl Sulfate–Polyacrylamide Gel Electrophoresis
Lyophilized
24.4kDa
E. coli expression system
25 ug
Store at -20°C. Reconstitute in water to a concentration of 1.0 mg/mL. Do not vortex. For extended storage, it is recommended to be further diluted in buffer containing carrier protein (e.g. 0.1% BSA) and stored in aliquots at -20°C to -80°C.
<0.1ng/μg (<1 EU/μg)
LIN28
Measured by its ability to induce fluorescence in Lin28 reporter cells (293 cells transfected with fluorescent protein genes under Lin28 control). OptimμM activity was achieved at 20μg/ml after incubation for 72 hr.
Escherichia coli
None
E. coli expression system
Lyophilized
79727
LIN28 (Human) Recombinant Protein
0.9
GPSVSNQQFAGGCAKAAEEAPEEAPEDAARAADEPQLLHGAGICKWFNVRMGFGFLSMTARAGVALDPPVDVFVHQSKLHMEGFRSLKEGEAVEFTFKKSAKGLESIRVTGPGGVFCIGSERRPKGKSMQKRRSKGDRCYNCGGLDHHAKECKLPPQPKKCHFCQSISHMVASCPLKAQQGP
RUO
CSDD1|FLJ12457|LIN-28|LIN28A|ZCCHC1
LIN28
Human ;Human
Yes