Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human LINS1 Full-length ORF (AAH10363.1, 1 a.a. - 436 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00055180P01S

 View more versions of this product

Catalog No. 89-950-725


Add to cart

Description

Description

The Drosophila segment polarity gene lin encodes a protein, lines, which plays important roles in development of the epidermis and hindgut. This gene encodes a protein containing a lines-like domain. This gene is located on chromosome 15 and clustered with the gene encoding ankyrin repeat and SOCS box-containing protein 7. Multiple transcript variants encoding the same protein have been found for this gene. [provided by RefSeq]

Sequence: MKVFCEVLEELYKKVLLGATLENDSHDYVFYLNPAVSDQDCSTATSLEWANTCGIQGRHQPISVGVAPIAVAPVCLKTNSQMSGSREVMLLQLTVIKVMTTRILSVKTEFHAKEQYRDVIKILLESAKVDSKLICMFQNSDKLLSHMAAQCLALLLYFQLREKITLSNSWIAFCQKNLSEYSESNKAIYCLWTLTAIIKEIFKDSCSQKTEILKQFLTHFDTIFEVFYNSLFSQHFENCRDTSKIVNILMCFLDLLELLIASRIHLKLHFTCQRILFLKPSCMLEVITWPIQAFVKRKVIIFLKKCLLCKVGEDLCRGSVPALMPPDHHVAVDMLALANAVLQAVNSGLLKTLSVYEKHSFFGGDEVQPECELITSPDHVILRAASLVIMKSLEIKFQNYSSASEMKGNSPNSFCMQCVIIYLSTVIHNYQISGLV
Specifications

Specifications

AAH10363.1
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
75.7kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FLJ10583|WINS1
LINS1
Yes
wheat germ expression system
Liquid
55180
LINS1 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MKVFCEVLEELYKKVLLGATLENDSHDYVFYLNPAVSDQDCSTATSLEWANTCGIQGRHQPISVGVAPIAVAPVCLKTNSQMSGSREVMLLQLTVIKVMTTRILSVKTEFHAKEQYRDVIKILLESAKVDSKLICMFQNSDKLLSHMAAQCLALLLYFQLREKITLSNSWIAFCQKNLSEYSESNKAIYCLWTLTAIIKEIFKDSCSQKTEILKQFLTHFDTIFEVFYNSLFSQHFENCRDTSKIVNILMCFLDLLELLIASRIHLKLHFTCQRILFLKPSCMLEVITWPIQAFVKRKVIIFLKKCLLCKVGEDLCRGSVPALMPPDHHVAVDMLALANAVLQAVNSGLLKTLSVYEKHSFFGGDEVQPECELITSPDHVILRAASLVIMKSLEIKFQNYSSASEMKGNSPNSFCMQCVIIYLSTVIHNYQISGLV
RUO
LINS1
Wheat Germ (in vitro)
GST
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit