Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human LMO1 Partial ORF (NP_002306.1, 15 a.a. - 156 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00004004Q02S

 View more versions of this product

Catalog No. 89-950-786


Add to cart

Description

Description

LMO1 enocdes a cysteine-rich, two LIM domain transcriptional regulator. It is mapped to an area of consistent chromosomal translocation in chromosome 11, disrupting it in T-cell leukemia, although more rarely than the related gene, LMO2 is disrupted. [provided by RefSeq]

Sequence: VQPKGKQKGCAGCNRKIKDRYLLKALDKYWHEDCLKCACCDCRLGEVGSTLYTKANLILCRRDYLRLFGTTGNCAACSKLIPAFEMVMRARDNVYHLDCFACQLCNQRFCVGDKFFLKNNMILCQMDYEEGQLNGTFESQVQ
Specifications

Specifications

NP_002306.1
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
41.25kDa
12.5% SDS-PAGE Stained with Coomassie Blue
VQPKGKQKGCAGCNRKIKDRYLLKALDKYWHEDCLKCACCDCRLGEVGSTLYTKANLILCRRDYLRLFGTTGNCAACSKLIPAFEMVMRARDNVYHLDCFACQLCNQRFCVGDKFFLKNNMILCQMDYEEGQLNGTFESQVQ
RUO
LMO1
Wheat Germ (in vitro)
GST
wheat germ expression system
Liquid
4004
LMO1 (Human) Recombinant Protein (Q02)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
MGC116692|RBTN1|RHOM1|TTG1
LMO1
Yes
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit