Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human LMO7 Partial ORF (NP_005349, 453 a.a. - 540 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00004008Q01S

 View more versions of this product

Catalog No. 89-950-793


Add to cart

Description

Description

This gene encodes a protein containing a calponin homology (CH) domain, a PDZ domain, and a LIM domain, and may be involved in protein-protein interactions. Several alternatively spliced transcript variants encoding different isoforms have been found for this gene, however, the full-length nature of some variants is not known. [provided by RefSeq]

Sequence: DRESQNQKSTVPSRRRMYSFDDVLEEGKRPPTMTVSEASYQSERVEEKGATYPSEIPKEDSTTFAKREDRVTTEIQLPSQSPVEEQSP
Specifications

Specifications

NP_005349
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
35.42kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
DRESQNQKSTVPSRRRMYSFDDVLEEGKRPPTMTVSEASYQSERVEEKGATYPSEIPKEDSTTFAKREDRVTTEIQLPSQSPVEEQSP
RUO
LMO7
Wheat Germ (in vitro)
GST
wheat germ expression system
Liquid
4008
LMO7 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FBX20|FBXO20|KIAA0858|LOMP
LMO7
Yes
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit