Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human LNX Partial ORF (NP_116011, 19 a.a. - 118 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00084708Q01L

 View more versions of this product

Catalog No. 89-950-811


Add to cart

Description

Description

This gene encodes a membrane-bound protein that is involved in signal transduction and protein interactions. The encoded product is an E3 ubiquitin-protein ligase, which mediates ubiquitination and subsequent proteasomal degradation of proteins containing phosphotyrosine binding (PTB) domains. This protein may play an important role in tumorogenesis. Alternatively spliced transcript variants encoding distinct isoforms have been described. A pseudogene, which is located on chromosome 17, has been identified for this gene. [provided by RefSeq]

Sequence: DNVGNLHFLYSELCKGAFHYGLTKDRKRRSQDGCPDGCASLTATAPSPEVSAAATISLMTDEPGLDNPAYVSSAEDGQPAISPVDSGRSNRTRARPFERS
Specifications

Specifications

NP_116011
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.74kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
DNVGNLHFLYSELCKGAFHYGLTKDRKRRSQDGCPDGCASLTATAPSPEVSAAATISLMTDEPGLDNPAYVSSAEDGQPAISPVDSGRSNRTRARPFERS
RUO
LNX1
Wheat Germ (in vitro)
GST
wheat germ expression system
Liquid
84708
LNX (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
LNX|MPDZ|PDZRN2
LNX1
Yes
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit