Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human LOC440295 Full-length ORF (AAH41854.1, 1 a.a. - 274 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00440295P01L

 View more versions of this product

Catalog No. 89-950-953


Add to cart

Description

Description

Sequence: MEQLQDETNHLRKELESVGRQLQAEVENNQMLSLLNRRQEERLREQEERLREQEERLCEQEERLCEQEERLREQEERLCEQEKLPGQERLLEEVEKLLEQERRQEEQERLLERERLLDEVEELLEQERLRQQDERLWQQETLRELERLRELERMLELGWEALYEQRAEPRSGFEELNNENKSTLQLEQQVKELEKSGGAEEPRGSESAAAARPVAGAPVPQGAWMCGQAGWTPQEHPGLSGEAVGTGEAAGGAGEAACHSFRAAENRELNITII
Specifications

Specifications

AAH41854.1
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
58.5kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
LOC440295
Wheat Germ (in vitro)
GST
wheat germ expression system
Liquid
440295
LOC440295 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MEQLQDETNHLRKELESVGRQLQAEVENNQMLSLLNRRQEERLREQEERLREQEERLCEQEERLCEQEERLREQEERLCEQEKLPGQERLLEEVEKLLEQERRQEEQERLLERERLLDEVEELLEQERLRQQDERLWQQETLRELERLRELERMLELGWEALYEQRAEPRSGFEELNNENKSTLQLEQQVKELEKSGGAEEPRGSESAAAARPVAGAPVPQGAWMCGQAGWTPQEHPGLSGEAVGTGEAAGGAGEAACHSFRAAENRELNITII
RUO
LOC440295
Yes
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit