Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human LOH11CR2A Full-length ORF (NP_055437.2, 1 a.a. - 786 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00004013P01S

 View more versions of this product

Catalog No. 89-951-019


Add to cart

Description

Description

Sequence: MVHFCGLLTLHREPVPLKSISVSVNIYEFVAGVSATLNYENEEKVPLEAFFVFPMDEDSAVYSFEALVDGKKIVAELQDKMKARTNYEKAISQGHQAFLLEGDSSSRDVFSCNVGNLQPGSKAAVTLKYVQELPLEADGALRFVLPAVLNPRYQFSGSSKDSCLNVKTPIVPVEDLPYTLSMVATIDSQHGIEKVQSNCPLSPTEYLGEDKTSAQVSLAAGHKFDRDVELLIYYNEVHTPSVVLEMGMPNMKPGHLMGDPSAMVSFYPNIPEDQPSNTCGEFIFLMDRSGSMQSPMSSQDTSQLRIQAAKETLILLLKSLPIGCYFNIYGFGSSYEACFPESVKYTQQTMEEALGRVKLMQADLGGTEILAPLQNIYRGPSIPGHPLQLFVFTDGEVTDTFSVIKEVRINRQKHRCFSFGIGEGTSTSLIKGIARASGGTSEFITGKDRMQSKALRTLKRSLQPVVEDVSLSWHLPPGLSAKMLSPEQTVIFRGQRLISYAQLTGRMPAAETTGEVCLKYTLQGKTFEDKVTFPLQPKPDVNLTIHRLAAKSLLQTKDMGLRETPASDKKDALNLSLESGVISSFTAFIAINKELNKPVQGPLAHRDVPRPILLGASAPLKIKCQSGFRKALHSDRPPSASQPRGELMCYKAKTFQMDDYSLCGLISHKDQHSPGFGENHLVQLIYHQNANGSWDLNEDLAKILGMSLEEIMAAQPAELVDSSGWATILAVIWLHSNGKDLKCEWELLERKAVAWMRAHAGSTMPSVVKAAITFLKSSVDPAIFAF
Specifications

Specifications

NP_055437.2
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
112.9kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
BCSC-1|BCSC1|LOH11CR2A
VWA5A
Yes
wheat germ expression system
Liquid
4013
LOH11CR2A (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MVHFCGLLTLHREPVPLKSISVSVNIYEFVAGVSATLNYENEEKVPLEAFFVFPMDEDSAVYSFEALVDGKKIVAELQDKMKARTNYEKAISQGHQAFLLEGDSSSRDVFSCNVGNLQPGSKAAVTLKYVQELPLEADGALRFVLPAVLNPRYQFSGSSKDSCLNVKTPIVPVEDLPYTLSMVATIDSQHGIEKVQSNCPLSPTEYLGEDKTSAQVSLAAGHKFDRDVELLIYYNEVHTPSVVLEMGMPNMKPGHLMGDPSAMVSFYPNIPEDQPSNTCGEFIFLMDRSGSMQSPMSSQDTSQLRIQAAKETLILLLKSLPIGCYFNIYGFGSSYEACFPESVKYTQQTMEEALGRVKLMQADLGGTEILAPLQNIYRGPSIPGHPLQLFVFTDGEVTDTFSVIKEVRINRQKHRCFSFGIGEGTSTSLIKGIARASGGTSEFITGKDRMQSKALRTLKRSLQPVVEDVSLSWHLPPGLSAKMLSPEQTVIFRGQRLISYAQLTGRMPAAETTGEVCLKYTLQGKTFEDKVTFPLQPKPDVNLTIHRLAAKSLLQTKDMGLRETPASDKKDALNLSLESGVISSFTAFIAINKELNKPVQGPLAHRDVPRPILLGASAPLKIKCQSGFRKALHSDRPPSASQPRGELMCYKAKTFQMDDYSLCGLISHKDQHSPGFGENHLVQLIYHQNANGSWDLNEDLAKILGMSLEEIMAAQPAELVDSSGWATILAVIWLHSNGKDLKCEWELLERKAVAWMRAHAGSTMPSVVKAAITFLKSSVDPAIFAF
RUO
VWA5A
Wheat Germ (in vitro)
GST
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit