Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human LPIN1 Full-length ORF (AAH30537, 1 a.a. - 890 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00023175P01

Catalog No. 89-951-047


Add to cart

Description

Description

This gene represents a candidate gene for human lipodystrophy, characterized by loss of body fat, fatty liver, hypertriglyceridemia, and insulin resistance. Mouse studies suggest that this gene functions during normal adipose tissue development and may also play a role in human triglyceride metabolism. [provided by RefSeq]

Sequence: MNYVGQLAGQVFVTVKGLYKGLNPATLSGCIDIIVIRQPNGNLQCSPFHVRFGKMGVLRSREKVVDIEINGESVDLHMKLGDNGEAFFVQETDNDQEVIPMHLATSPILSEGASRMECQLKRGSVDRMRGLDPSTPAQVIAPSETPSSSSVVKKRRKRRRKSQLDSLKRDDNMNTSEDEDMFPIEMSSDEAMELLESSRTLPNDIPPFQDDIPEENLSLAVIYPQSASYPNSDREWSPTPSPSGSRPSTPKSDSELVSKSTERTGQKNPEMLWLWGELPQAAKSSSPHKMKESSPLSSRKICDKSHFQAIHSESSDTFSDQSPTLVGGALLDQNKPQTEMQFVNEEDLETLGAAAPLLPMIEELKPPSASVVQTANKTDSPSRKRDKRSRHLGADGVYLDDLTDMDPEVAALYFPKNGDPSGLAKHASDNGARSANQSPQSVGSSGVDSGVESTSDGLRDLPSIAISLCGGLSDHREITKDAFLEQAVSYQQFVDNPAIIDDPNLVVKIGSKYYNWTTAAPLLLAMQAFQKPLPKATVESIMRDKMPKKGGRWWFSWRGRNTTIKEESKPEQCLAGKAHSTGEQPPQLSLATRVKHESSSSDEERAAAKPSNAGHLPLLPNVSYKKTLRLTSEQLKTLKLKNGPNDVVFSVTTQYQGTCRCEGTIYLWNWDDKVIISDIDGTITRSDTLGHILPTLGKDWTHQGIAKLYHKVSQNGYKFLYCSARAIGMADMTRGYLHWVNERGTVLPQGPLLLSPSSLFSALHREVIEKKPEKFKVQCLTDIKNLFFPNTEPFYAAFGNRPADVYSYKQVGVSLNRIFTVNPKGELVQEHAKTNISSYVRLCEVVDHVFPLLKRSHSSDFPCSDTFSNFTFWREPLPPFENQDIHSASA
Specifications

Specifications

AAH30537
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
123.64kDa
Glutathione Sepharose 4 Fast Flow
2 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
DKFZp781P1796|KIAA0188|PAP1
LPIN1
Yes
wheat germ expression system
Liquid
23175
LPIN1 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MNYVGQLAGQVFVTVKGLYKGLNPATLSGCIDIIVIRQPNGNLQCSPFHVRFGKMGVLRSREKVVDIEINGESVDLHMKLGDNGEAFFVQETDNDQEVIPMHLATSPILSEGASRMECQLKRGSVDRMRGLDPSTPAQVIAPSETPSSSSVVKKRRKRRRKSQLDSLKRDDNMNTSEDEDMFPIEMSSDEAMELLESSRTLPNDIPPFQDDIPEENLSLAVIYPQSASYPNSDREWSPTPSPSGSRPSTPKSDSELVSKSTERTGQKNPEMLWLWGELPQAAKSSSPHKMKESSPLSSRKICDKSHFQAIHSESSDTFSDQSPTLVGGALLDQNKPQTEMQFVNEEDLETLGAAAPLLPMIEELKPPSASVVQTANKTDSPSRKRDKRSRHLGADGVYLDDLTDMDPEVAALYFPKNGDPSGLAKHASDNGARSANQSPQSVGSSGVDSGVESTSDGLRDLPSIAISLCGGLSDHREITKDAFLEQAVSYQQFVDNPAIIDDPNLVVKIGSKYYNWTTAAPLLLAMQAFQKPLPKATVESIMRDKMPKKGGRWWFSWRGRNTTIKEESKPEQCLAGKAHSTGEQPPQLSLATRVKHESSSSDEERAAAKPSNAGHLPLLPNVSYKKTLRLTSEQLKTLKLKNGPNDVVFSVTTQYQGTCRCEGTIYLWNWDDKVIISDIDGTITRSDTLGHILPTLGKDWTHQGIAKLYHKVSQNGYKFLYCSARAIGMADMTRGYLHWVNERGTVLPQGPLLLSPSSLFSALHREVIEKKPEKFKVQCLTDIKNLFFPNTEPFYAAFGNRPADVYSYKQVGVSLNRIFTVNPKGELVQEHAKTNISSYVRLCEVVDHVFPLLKRSHSSDFPCSDTFSNFTFWREPLPPFENQDIHSASA
RUO
LPIN1
Wheat Germ (in vitro)
GST
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit