Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abnova Human LRRC23 Full-length ORF (AAH29858.1, 1 a.a. - 343 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
$416.00 - $641.00
Specifications
Name | LRRC23 (Human) Recombinant Protein (P01) |
---|---|
Recombinant | Yes |
Purification Method | Glutathione Sepharose 4 Fast Flow |
For Use With (Application) | Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein) |
Molecular Weight | 66.1kDa |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
89-951-112
|
Abnova
H00010233P01L |
25 ug |
Each for $641.00
|
|
89-951-111
|
Abnova
H00010233P01S |
10 ug |
Each for $416.00
|
|
Description
Sequence: MSDEDDLEDSEPDQDDSEKEEDEKETEEGEDYRKEGEEFPEEWLPTPLTEDMMKEGLSLLCKTGNGLAHAYVKLEVKERDLTDIYLLRSYIHLRYVDISENHLTDLSPLNYLTHLLWLKADGNLLRSAQMNELPYLQIASFAYNQITDTEGISHPRLETLNLKGNSIHMVTGLDPEKLISLHTVELRGNQLESTLGINLPKLKNLYLAQNMLKKVEGLEDLSNLTTLHLRDNQIDTLSGFSREMKSLQYLNLRGNMVANLGELAKLRDLPKLRALVLLDNPCTDETSYRQEALVQMPYLERLDKEFYEEEERAEADVIRQRLKEEKEQEPEPQRDLEPEQSLISpecifications
LRRC23 (Human) Recombinant Protein (P01) | |
Glutathione Sepharose 4 Fast Flow | |
66.1kDa |
Yes | |
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein) | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title