Learn More
Abnova™ Human LRRC29 Partial ORF (NP_036295, 125 a.a. - 223 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Supplier: Abnova™ H00026231Q01L
Description
This gene encodes a member of the F-box protein family which is characterized by an approximately 40 amino acid motif, the F-box. The F-box proteins constitute one of the four subunits of ubiquitin protein ligase complex called SCFs (SKP1-cullin-F-box), which function in phosphorylation-dependent ubiquitination. The F-box proteins are divided into 3 classes: Fbws containing WD-40 domains, Fbls containing leucine-rich repeats, and Fbxs containing either different protein-protein interaction modules or no recognizable motifs. The protein encoded by this gene belongs to the Fbls class and, in addition to an F-box, contains 9 tandem leucine-rich repeats. Two transcript variants encoding the same protein have been found for this gene. Other variants may occur, but their full-length natures have not been characterized. [provided by RefSeq]
Sequence: CPSLEHLALSHCSRLSDKGWAQAASSWPRLQHLNLSSCSQLIEQTLDAIGQACRQLRVLDVATCPGINMAAVRRFQAQLPQVSCVQSRFVGGADLTLTLSpecifications
NP_036295 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.63kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
CPSLEHLALSHCSRLSDKGWAQAASSWPRLQHLNLSSCSQLIEQTLDAIGQACRQLRVLDVATCPGINMAAVRRFQAQLPQVSCVQSRFVGGADLTLTL | |
RUO | |
LRRC29 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
26231 | |
LRRC29 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FBL9/FBXL9 | |
LRRC29 | |
Recombinant | |
wheat germ expression system |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.