Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems™ Recombinant Human LRRC4 Fc Chimera Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.
Supplier: R&D Systems™ 9375LR050
This item is not returnable.
View return policy
Description
The Recombinant Human LRRC4 Fc Chimera Protein is derived from HEK293. The Recombinant Human LRRC4 Fc Chimera Protein has been validated for the following applications: Bioactivity.Order Info
Ships at ambient temperature.
Specifications
Q9HBW1 | |
Lyophilized from a 0.2μm filtered solution in PBS | |
81 kDa | |
Human embryonic kidney cell,HEK293-derived human LRRC4 protein, (N-terminus) Human LRRC4 (Ala39-Lys527) Accession &Num; Q9HBW1 IEGRMD Human IgG1 (Pro100-Lys330) (C-terminus) | |
RUO | |
BAG, Brain Tumor Associated Protein LRRC4, Brain Tumor-Associated Protein BAG | |
Recombinant protein | |
Human | |
>95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining |
Bioactivity | |
64101 | |
50μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
<0.10 EU per 1μg of the protein by the LAL method. | |
2 to 12ng/mL | |
Unconjugated | |
Recombinant |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only.
Spot an opportunity for improvement?
Provide Content Correction