Learn More
Abnova™ Human LSM1 Partial ORF (NP_055277.1, 34 a.a. - 133 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Manufacturer: Abnova™ H00027257Q01L
Description
Sm-like proteins were identified in a variety of organisms based on sequence homology with the Sm protein family (see SNRPD2; MIM 601061). Sm-like proteins contain the Sm sequence motif, which consists of 2 regions separated by a linker of variable length that folds as a loop. The Sm-like proteins are thought to form a stable heteromer present in tri-snRNP particles, which are important for pre-mRNA splicing.[supplied by OMIM]
Sequence: SIDQFANLVLHQTVERIHVGKKYGDIPRGIFVVRGENVVLLGEIDLEKESDTPLQQVSIEEILEEQRVEQQTKLEAEKLKVQALKDRGLSIPRADTLDEYSpecifications
NP_055277.1 | |
wheat germ expression system | |
Liquid | |
CASM|YJL124C | |
LSM1 | |
SIDQFANLVLHQTVERIHVGKKYGDIPRGIFVVRGENVVLLGEIDLEKESDTPLQQVSIEEILEEQRVEQQTKLEAEKLKVQALKDRGLSIPRADTLDEY | |
LSM1 (Human) Recombinant Protein (Q01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Yes | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
LSM1 | |
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
27257 | |
Wheat Germ (in vitro) | |
36.74kDa | |
GST | |
25 ug | |
RUO |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.