Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human LSM4 Partial ORF (NP_036453.1, 1 a.a. - 73 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00025804Q01L

 View more versions of this product

Catalog No. 89-951-212


Add to cart

Description

Description

Sm-like proteins were identified in a variety of organisms based on sequence homology with the Sm protein family (see SNRPD2; MIM 601061). Sm-like proteins contain the Sm sequence motif, which consists of 2 regions separated by a linker of variable length that folds as a loop. The Sm-like proteins are thought to form a stable heteromer present in tri-snRNP particles, which are important for pre-mRNA splicing.[supplied by OMIM]

Sequence: MLPLSLLKTAQNHPMLVELKNGETYNGHLVSCDNWMNINLREVICTSRDGDKFWRMPECYIRGSTIKYLRIPD
Specifications

Specifications

wheat germ expression system
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
33.77kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
MLPLSLLKTAQNHPMLVELKNGETYNGHLVSCDNWMNINLREVICTSRDGDKFWRMPECYIRGSTIKYLRIPD
RUO
LSM4
Wheat Germ (in vitro)
GST
NP_036453.1
Liquid
25804
LSM4 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
YER112W
LSM4
Yes
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit