Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human LSS Partial ORF (NP_001001438.1, 633 a.a. - 732 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00004047Q01S

 View more versions of this product

Catalog No. 89-951-223


Add to cart

Description

Description

The protein encoded by this gene catalyzes the conversion of (S)-2,3 oxidosqualene to lanosterol. The encoded protein is a member of the terpene cyclase/mutase family and catalyzes the first step in the biosynthesis of cholesterol, steroid hormones, and vitamin D. Alternative splicing results in multiple transcript variants encoding different isoforms

Sequence: FESCEERRYLQSAQSQIHNTCWAMMGLMAVRHPDIEAQERGVRCLLEKQLPNGDWPQENIAGVFNKSCAISYTSYRNIFPIWALGRFSQLYPERALAGHP
Specifications

Specifications

NP_001001438.1
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.74kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
FESCEERRYLQSAQSQIHNTCWAMMGLMAVRHPDIEAQERGVRCLLEKQLPNGDWPQENIAGVFNKSCAISYTSYRNIFPIWALGRFSQLYPERALAGHP
RUO
LSS
Wheat Germ (in vitro)
GST
wheat germ expression system
Liquid
4047
LSS (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FLJ25486|FLJ35015|FLJ39450|FLJ46393|OSC
LSS
Yes
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit