Learn More
Abnova™ Human LSS Partial ORF (NP_001001438.1, 633 a.a. - 732 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
$499.00 - $769.00
Specifications
Accession Number | NP_001001438.1 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 4047 |
Molecular Weight (g/mol) | 36.74kDa |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
89-951-224
|
Abnova™
H00004047Q01L |
25 ug |
Each for $769.00
|
|
|||||
89-951-223
|
Abnova™
H00004047Q01S |
10 ug |
Each for $499.00
|
|
|||||
Description
The protein encoded by this gene catalyzes the conversion of (S)-2,3 oxidosqualene to lanosterol. The encoded protein is a member of the terpene cyclase/mutase family and catalyzes the first step in the biosynthesis of cholesterol, steroid hormones, and vitamin D. Alternative splicing results in multiple transcript variants encoding different isoforms
Sequence: FESCEERRYLQSAQSQIHNTCWAMMGLMAVRHPDIEAQERGVRCLLEKQLPNGDWPQENIAGVFNKSCAISYTSYRNIFPIWALGRFSQLYPERALAGHPSpecifications
NP_001001438.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ25486/FLJ35015/FLJ39450/FLJ46393/OSC | |
LSS | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
4047 | |
LSS (Human) Recombinant Protein (Q01) | |
FESCEERRYLQSAQSQIHNTCWAMMGLMAVRHPDIEAQERGVRCLLEKQLPNGDWPQENIAGVFNKSCAISYTSYRNIFPIWALGRFSQLYPERALAGHP | |
RUO | |
LSS | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.