Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human LTB4DH Partial ORF (NP_036344, 230 a.a. - 328 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00022949Q01L

 View more versions of this product

Catalog No. 89-951-241


Add to cart

Description

Description

This gene encodes an enzyme that is involved in the inactivation of the chemotactic factor, leukotriene B4. The encoded protein specifically catalyzes the NADP+ dependent conversion of leukotriene B4 to 12-oxo-leukotriene B4. A pseudogene of this gene is found on chromosome 1. Alternative splicing results in multiple transcript variants. [provided by RefSeq]

Sequence: MKKFGRIAICGAISTYNRTGPLPPGPPPEIVIYQELRMEAFVVYRWQGDARQKALKDLLKWVLEGKIQYKEYIIEGFENMPAAFMGMLKGDNLGKTIVK
Specifications

Specifications

wheat germ expression system
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.63kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
MKKFGRIAICGAISTYNRTGPLPPGPPPEIVIYQELRMEAFVVYRWQGDARQKALKDLLKWVLEGKIQYKEYIIEGFENMPAAFMGMLKGDNLGKTIVK
RUO
PTGR1
Wheat Germ (in vitro)
GST
NP_036344
Liquid
22949
LTB4DH (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
LTB4DH/MGC34943/ZADH3
PTGR1
Yes
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit