Learn More
Abnova™ Human LTK (NP_002335.2, 498 a.a. - 796 a.a.) Partial Recombinant Protein with GST-tag expressed in baculovirus infected Sf21 cells
Used for Func, SDS-PAGE
Manufacturer:  Abnova™ P5581
Catalog No. 89-951-251
Add to cart
Description
Description
The protein encoded by this gene is a member of the ros/insulin receptor family of tyrosine kinases. Tyrosine-specific phosphorylation of proteins is a key to the control of diverse pathways leading to cell growth and differentiation. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]
Specifications
Specifications
NP_002335.2
Functional Study, Sodium Dodecyl Sulfate–Polyacrylamide Gel Electrophoresis
Liquid
60kDa
Baculovirus infected insect cell (Sf21) expression system
87% by SDS-PAGE/CBB staining
5 ug
RUO
LTK
The activity was measured by off-chip mobility shift assay. The enzyme was incubated with fluorescence-labeled substrate and Mg(or Mn)/ATP. The phosphorylated and unphosphorylated substrates were separated and detected by LabChip 3000. Substrate: Srctide. ATP: 100 uM.
Yes
Baculovirus infected insect cell (Sf21) expression system
Liquid
4058
LTK (Human) Recombinant Protein
Glutathione sepharose chromatography
Loading 1 ug protein in SDS-PAGE
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
TYK1
LTK
Insect
GST