Learn More
Raybiotech IncSupplier Diversity Partner Human Lumican ELISA Kit DFS Item
Manufacturer:  Raybiotech IncSupplier Diversity Partner ELHLUM1
For Cell Culture Supernatants, Plasma, and Serum samples. Sensitivity: 0.1 ng/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair recognizes Human Lumican. Other species not yet determined.
Catalog No. 50-194-9515